missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Hubtron™ Advance Fluoroelastomer Tubing

Ideal for corrosive chemical transfer, electronic applications, ink and general printing applications, and automotive applications

Brand:  Cole-Parmer™ 95226-00

 View more versions of this product

Product Code. 11785298

  • £595.00 / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Transparent, very low gas permeability, highly chemical compatible.
  • Inner fluoroplastic liner, outer fluoroelastomer cover.
  • Smooth inner lining.
  • Excellent alcohol, ester, acid and alkali resistance.
  • Autoclavable.
Specifications

Specifications

Resin: FDA;Non-DEHP;Non-Phthalate;European Pharmacopoeia;NSF-61;NSF-51;3A;ADCF;USP
1.6 mm
4.8 mm
7.6 m
14°F
Transparent
120°C
Autoclavable
PTFE
25 ft.
1.6 mm
-10°C
248°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers