missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human 20S Proteasome X (aa 243-259) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP172
This item is not returnable.
View return policy
Description
Human 20S Proteasome X (residues 243-259) Synthetic Peptide, PEP-172, from Invitrogen.
PEP-172 is an 18 amino acid synthetic peptide whose sequence is derived from human proteasome 20S X protein. The sequence of this peptide is (amino to carboxy terminus): C-W (243)-I-R-V-S-S-D-N-V-A-D-L-H-D-K-Y-S (259)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human proteasome 20S X, catalog # PA1-977. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
5693 | |
purified | |
Human 20S Proteasome X (aa 243-259) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P28074 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
PSMB5 | |
No preservative | |
50 μg | |
C-W(243)-I-R-V-S-S-D-N-V-A-D-L-H-D-K-Y-S(259) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction