missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM22 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4908-SP
This item is not returnable.
View return policy
Description
ADAM22 Polyclonal specifically detects ADAM22 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ADAM22 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ADAM 22, ADAM metallopeptidase domain 22, MDC2, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832 | |
Sheep | |
Affinity Purified | |
RUO | |
53616 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q9P0K1 | |
ADAM22 | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAM22 Gly26-His528 (Leu78Val) Accession # Q9P0K1 | |
25 μg | |
Primary | |
Detects human ADAM22 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ADAM8, rhADAM12, and rhADAM33 is observed. | |
Human | |
IgG |
For Research Use Only