missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ADAMTS5 Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP77529
This item is not returnable.
View return policy
Description
ADAMTS5 (human) Recombinant Protein, RP-77529, from Invitrogen.
RP-77529 is a purified recombinant human ADAMTS5 D625-930, produced with the baculovirus expression system and purified from insect cell culture supernatants. The protein contains the catalytic domain, the disintegrin domain and the thrombospondin type-1 motif of full-lenth ADAMTS5 followed by a C-terminal His6-tag. The calculated MW of the amino acid sequence is 40.8 kDa. In SDS-PAGE the protein exhibits a MW of about 50 kDa. The higher Mr is probably due to glycosylation. RP-77529 may be used as a control with corresponding anti-ADAMTS5 antibodies.Specifications
Q9UNA0 | |
Control | |
11096 | |
7.5 | |
Insect Cells | |
RUO | |
ADAMTS5 | |
Unconjugated | |
Baculovirus | |
Liquid | |
purified |
0.1mg/mL | |
TBS with 0.05% Brij-35, 50mM Imidazole, 5mM CaCl2 and no preservative; pH 7.5 | |
Human ADAMTS5 | |
5 μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
Human ADAMTS5 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro research only.