missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human Albumin ELISA Kit

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous albumin

Brand:  Proteintech™ KE00076-96T

208.05 GBP valid until 2024-12-31
Use promo code "21957" to get your promotional price.



Product Code. 16801496

  • £219.00 / 96 tests
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Recognizes human albumin
  • Albumin most abundant protein in blood plasma
    • Produced by well-differentiated hepatocytes
    • Plays role in maintaining osmotic pressure
    • Decreased levels of serum albumin may be sign of kidney or liver diseases and diabetes

Process:

  • Antibody specific for Albumin pre-coated onto microwells
  • Albumin protein in samples captured by the coated antibody after incubation
  • After extensive washing, another horseradish peroxidase (HRP)-conjugated antibody specific for Albumin is added to detect captured Albumin
  • For signal development, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications

Specifications

NP_000468
12 ng/mL
Sandwich ELISA, Double Antibody
Plasma, Serum
Human
Human
<10%
Blue ice
2.5 hr.
50 to 3200 ng/mL
ISO 13485
Coated ELISA Kit
DAB
213
Solid
<10%
-20°C to +4°C
P02768
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 96TEST Human PG ELISA Kit

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers