missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-Synuclein Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB11376-100
This item is not returnable.
View return policy
Description
alpha-Synuclein Monoclonal antibody specifically detects alpha-Synuclein in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry
Specifications
| alpha-Synuclein | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), Synuclein-alpha, truncated alpha synuclein | |
| E. coli -derived human alpha-Synuclein, Met1-Ala140, Accession # P37840 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry, Immunocytochemistry | |
| 1060506 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| P37840 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6622 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction