missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Amyloid Precursor Protein Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0826
Code : NEW
Additional Details : Weight : 0.01000kg
Description
APP Synthetic Peptide, PEP-0826, from Invitrogen.
Amyloid Precursor Protein (APP) or Amyloid beta protein functions as a cell surface kinesin I membrane receptor, mediating the axonal transport of beta-secretase and presenilin 1; APP is rapidly internalized via clathrin-coated pits. APP is important for neurite growth, neuronal adhesion and axonogenesis. Mutations in the APP gene can lead to Alzheimer disease 1 (AD1). This anitbody recognizes APP (b-APP770) with a MW of approximately 100kDa. This peptide corresponds to 18 amino acids near the amino terminus of human APP.PEP-0826 can be used as a blocking peptide with polyclonal antibody PA5-20711.Specifications
351 | |
purified | |
Human Amyloid Precursor Protein | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
P05067 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
APP | |
0.02% sodium azide | |
50 μg | |
PBS |