missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human Apo-SAA1, Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ AF-300-53-10UG
71.55 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
The serum amyloid A family comprises a number of differentially expressed apolipoproteins, acute-phase SAA1 and SAA2, the former being the major component in plasma and constitutive SAAs. Although the liver is the primary site of synthesis of both SAA types extrahepatic production has been reported. The in vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed its importance in the diagnosis and monitoring of various diseases. Pathological SAA values are often detected in association with normal CRP concentrations; SAA rises earlier and more sharply than CRP. Recently, a broader view of SAA expression and function has been emerging.Specifications
P0DJI8 | |
protein with no preservative | |
11.7 | |
10 μg | |
-20°C | |
<0.1 EU/μg | |
SAA1 | |
Unconjugated | |
MRSFFSFLGE AFDGARDMWR AYSDMREANY IGSDKYFHAR GNYDAAKRGP GGVWAAEAIS DARENIQRFF GHGAEDSLAD QAANEWGRSG KDPNHFRPAG LPEKY | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
6288 | |
Human Apo-SAA1, Animal-Free | |
E. coli | |
RUO | |
amyloid a; Amyloid fibril protein AA; Amyloid protein A; MGC111216; PIG4; SAA; SAA1; Saa-1; Saa2; serum amyloid A; serum amyloid A 1; serum amyloid A 2; serum amyloid A protein; serum amyloid A1; Serum amyloid A1 protein; Serum amyloid A-1 protein; serum amyloid A-like protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); Serum Amyloid-A; TP53I4; tumor protein p53 inducible protein 4 | |
Human | |
Recombinant | |
Lyophilized |