missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BACE1 (aa 485-501) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP093
This item is not returnable.
View return policy
Description
Human beta-secretase (residues 485-501) Synthetic Peptide, PEP-093, from Invitrogen.
PEP-093 is a 16 amino acid synthetic peptide whose sequence corresponds to residues 485-501 of human beta-secretase conjugated to KLH. The sequence of this peptide is (amino to carboxy terminus): C485-L-R-Q-Q-H-D-D-F-A-D-D-I-S-L-L-K501This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human beta-secretase, catalog # PA1-757. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
23621 | |
purified | |
Human BACE1 (aa 485-501) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P56817 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
BACE1 | |
No preservative | |
50 μg | |
C485 - L - R - Q - Q - H - D - D - F - A - D - D - I - S - L - L - K501 |