missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BCRP Vesicles

Human BCRP Vesicles are insect derived purified plasma membranes with inserted BCRP transport protein (Breast Cancer Resistance Protein, ABCG2). Our Human BCRP Vesicles can be used to evaluate test compounds and drug interactions with BCRP in in vitro assays.

Brand:  Invitrogen™ GM0008

Product Code. 10589900

  • £1188.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Frozen
Research Grade
Human Vesicle
Vial
0.5 mL
Human
Store in ultracold freezer (-60 to -85°C) and protect from light.
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberKSAOG23 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberGM0008
Lot NumberBSH1G29 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberGM0008
Lot NumberGMUOG33 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberGM0008
Lot NumberJ6EL403 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberGM0008
Lot NumberBKAPG29 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberGM0008
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Human BCRP Vesicles > BCRP Vesicles

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.