Learn More
Invitrogen™ Human beta-Arrestin 2 (aa 384-397) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP156
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human beta Arrestin2 (residues 384-397) Synthetic Peptide, PEP-156, from Invitrogen.
PEP-156 is a 14 amino acid synthetic peptide whose sequence is derived from human beta-arrestin2 protein. The sequence of this peptide is (amino to carboxy terminus): C-D (384)-D-I-V-F-E-D-F-A-R-L-R-L-K (397)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human beta-arrestin2, catalog # PA1-730. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
409 | |
purified | |
Human beta-Arrestin 2 (aa 384-397) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P32121 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
ARRB2 | |
No preservative | |
50 μg | |
C-D(384)-D-I-V-F-E-D-F-A-R-L-R-L-K(397) |