Learn More
Novus Biologicals™ Human beta-Galactosidase-1/GLB1 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand: Novus Biologicals™ NBP3-18790
Additional Details : Weight : 0.00970kg
Specifications
3.13-200 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human beta-Galactosidase-1/GLB1 in samples. No significant cross-reactivity or interference between Human beta-Galactosidase-1/GLB1 and analogues was observed. | |
2720 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Human |
1.88 pg/mL | |
Biological Fluids, Plasma, Serum | |
ELISA, Sandwich ELISA | |
GLB1 | |
metabolism | |
Acid beta-galactosidase, beta-galactosidase, EBP, EC 3.2.1.23, Elastin receptor 1, elastin receptor 1 (67kD), elastin receptor 1, 67kDa, ELNR1, galactosidase, beta 1, Lactase, MPS4B |