missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BMP-4 Recombinant Protein
Recombinant Human Bone Morphogenetic Protein 4 (BMP4) is a bioactive protein intended for use in cell culture applications
£219.00 - £6690.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene Alias | BMP-4 |
Species | Human |
Shipping Condition | Dry Ice |
Expression System | Mammalian |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11330852
|
Gibco™
PHC9534 |
10μg |
£219.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11310852
|
Gibco™
PHC9531 |
100μg |
£2085.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320852
|
Gibco™
PHC9533 |
1mg |
£6690.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human BMP-4 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
BMP4 is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by its ability to induce alkaline phosphatase production by ATDC-5 cells
Specifications
Cell Culture | |
Human | |
Mammalian | |
Recombinant, Ligand | |
BMPs (Bone Morphogenetic Proteins) & Related Factors | |
Gibco™ |
BMP-4 | |
Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Bone Research, Stem Cell Research, Differentiation |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.