missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BMP-4 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 120-05ET-250UG
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.Specifications
P12644 | |
protein with no preservative | |
23.9 | |
250 μg | |
-20°C | |
<1 EU/μg | |
BMP4 | |
Unconjugated | |
KKNKNCRRHS LYVDFSDVGW NDWIVAPPGY QAFYCHGDCP FPLADHLNST NHAIVQTLVN SVNSSIPKAC CVPTELSAIS MLYLDEYDKV VLKNYQEMVV EGCGCR | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
652 | |
Human BMP-4 | |
E. coli | |
RUO | |
BMP; BMP2B; BMP-2 B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2 B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME | |
Human | |
Recombinant | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction