Learn More
Novus Biologicals™ Human Brain Occipital Lobe Whole Tissue Lysate (Adult Whole Parkinson's)
Brand: Novus Biologicals™ NB820-59413
Additional Details : Weight : 0.00970kg
Description
The Human Brain Occipital Lobe Whole Tissue Lysate (Adult Whole Parkinson's) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.Specifications
Western Blot | |
1 mg | |
Total Protein - Parkinsons Disease: Brain: Occipital Lobe | |
Parkinson's | |
Human | |
Human Brain Occipital Lobe Whole Tissue Lysate (Adult Parkinson's) | |
Adult |
Nervous | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Whole | |
Human | |
5 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
For Research Use Only