Learn More
Invitrogen™ Human BSEP Vesicles
Human BSEP Vesicles are insect derived purified plasma membranes with inserted BSEP transport protein (bile salt export pump, ABCB11). Our Human BSEP Vesicles can be used to evaluate test compounds and drug interactions with BSEP in in vitro assays.
Brand:  Invitrogen™ GM0005
Product Code. 10308189
  • £994.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Specifications
Frozen
Research Grade
Human Vesicle
Vial
0.5 mL
Human
Store at -80°C.
Dry Ice