missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Carbonic Anhydrase II/CA2 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF2184
Additional Details : Weight : 0.00970kg
Description
Carbonic Anhydrase II/CA2 Polyclonal specifically detects Carbonic Anhydrase II/CA2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation.Specifications
Carbonic Anhydrase II/CA2 | |
Polyclonal | |
Unconjugated | |
P00918 | |
CA2 | |
E. coli-derived recombinant human Carbonic Anhydrase II/CA2 Ser2-Lys260 Accession # P00918 | |
100 μg | |
Primary | |
Detects human Carbonic Anhydrase II/CA2 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Carbonic Anhydrase I, III, and XIII is observed and less than 1% cross-reactivity with rhCarbonic Anhydrase IV, VIII, IX, X, XII, and XIV is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
CA2, CAII, CA-IIEC 4.2.1.1, Car2, Carbonate dehydratase II, carbonic anhydrase 2, carbonic anhydrase B, Carbonic anhydrase C, carbonic anhydrase IICAC, carbonic dehydratase | |
Sheep | |
Affinity Purified | |
RUO | |
760 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |