Learn More
Invitrogen™ Human Carbonic Anhydrase X (aa 126-202) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP101386
Product Code. 30211991
  • £225.00 / 100µL
Explore more special offers
Description
Description
This gene encodes a protein that belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. The protein encoded by this gene is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development. Multiple transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q9NS85
Blocking Assay, Control
56934
100 μL
RUO
CA10
Human
HRLEEIRLHFGSEDSQGSEHLLNGQAFSGEVQLIHYNHELYTNVTEAAKSPNGLVVVSIFIKVSDSSNPFLNRMLNR
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
Carbonic Anhydrase X
-20° C, Avoid Freeze/Thaw Cycles
2700029L05Rik; BB085816; Ca10; Car10; carbonic anhydrase 10; carbonic anhydrase X; carbonic anhydrase-related protein 10; Carbonic anhydrase-related protein X; CARP X; CA-RP X; CARPX; CA-RPX; Cerebral protein 15; cerebral protein-15; HUCEP-15; UNQ533/PRO1076
Unconjugated
Recombinant
E. coli