missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Carboxypeptidase B2 Native Protein
Native Protein
Brand: Invitrogen™ RP43139
This item is not returnable.
View return policy
Description
Thrombin Activ. Fibrinolysis Inhib. (human) Native Protein, RP-43139, from Invitrogen.
RP-43139 contains native human Thrombin Activatable Fibrinolysis Inhibitor protein. RP-43139 may be used as a positive control in many immunological assays. The RP-43139 protein molecular weight is ∽60 kDa. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!Specifications
Q96IY4 | |
Control | |
1361 | |
Human Carboxypeptidase B2 | |
50μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
Human Carboxypeptidase B2 | |
Human | |
Hu | |
Native | |
Native |
2.5mg/mL | |
0.02M HEPES with 0.15M NaCl and no preservative; pH 7.4 | |
60 | |
7.4 | |
Human | |
RUO | |
CPB2 | |
Unconjugated | |
Human | |
Liquid | |
purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
in vitro research only.
Spot an opportunity for improvement?Share a Content Correction