missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Human Cardiomyocyte Immunocytochemistry Kit

Human Cardiomyocyte Immunocytochemistry Kit

Brand:  Molecular Probes™ A25973

Product Code. 15307695

  • £422.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2 Color, Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594

Store at -20°C to 4°C:

  • 10 μL of Mouse Anti-Human TNNT2 (1000X)
  • 10 μL Rabbit Anti-Human NKX2.5 (1000x)
  • 40 μL of Alexa Fluor™ 555 Donkey Anti-Rabbit (250X)
  • 40 μL of Alexa Fluor™ 594 Donkey Anti-Rabbit (250X)
  • 40 μL of Alexa Fluor™ 488 Donkey Anti-Mouse(250X)

Store at -20°C to RT:

  • 1 vial of NucBlue™ Fixed Cell ReadyProbes™ Reagent (1X)
  • 10 mL of Fixative Solution (1X)
  • 10 mL of Permeabilization Solution S (1X)
  • 20 mL of Blocking Solution (1X)
  • 20 mL of Wash Buffer (10X)
Immunocytochemistry Kit
50 tests
Alexa Fluor™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3185104 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberA25973
Lot Number3175174 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberA25973
Lot Number3142853 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberA25973
Lot Number2963807 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberA25973
Lot Number2963792 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberA25973
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Human Cardiomyocyte Immunocytochemistry Kit > 50 tests

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.