missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Caspase-1 ELISA Kit
Human Caspase-1 quantitates human Caspase-1 in serum, plasma, supernatant.
£540.00 - £2320.00
Specifications
Accession Number | CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1 B converting enzyme; IL1BC; IL-1 BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1 beta converting enzyme; interleukin-1 beta convertase; interleukin-1 beta-converting enzyme; p45 |
---|---|
Assay Range | 0.43 to 100 ng/mL |
Assay Sensitivity | 0.43 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17195299
|
Invitrogen™
EH70RBX5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
|||||
16895441
|
Invitrogen™
EH70RB |
96 Tests |
£540.00
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
Description
Human Caspase-1 quantitates human Caspase-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Caspase-1.
Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12 and can activate proinflammatory cytokines such as IL-1-b and IL-18. Caspase-1 was initially identified as an IL-1-b-converting enzyme; later experiments revealed it to be a mammalian homolog of the C. elegans cell death gene ced-3 whose overexpression can induce apoptosis in fibroblasts.Specifications
CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1 B converting enzyme; IL1BC; IL-1 BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1 beta converting enzyme; interleukin-1 beta convertase; interleukin-1 beta-converting enzyme; p45 | |
0.43 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CASP1 | |
<10% | |
HRP | |
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL | |
Caspase 1 | |
4 hr 45 min |
0.43 to 100 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P29466 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |