missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Cathepsin C ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EH73RBX10
Additional Details : Weight : 2.50000kg
Description
Cathepsin C, known also as dipeptidyl aminopeptidase I (DPPI), is a tetrameric lysosomal cysteine peptidase belonging to the papain family. Cathepsin C is involved in intracellular protein degradation and the processing of protein precursors, where it participates in cell growth, neuraminidase activation, and platelet factor XIII activation. Cathepsin C is largely related to other lysosomal cysteine proteinases, including cathepsin B, H and L. Enzymatically, Cathepsin C is capable of sequentially removing dipeptides from the amino terminus, and it requires halide ions, namely chloride ions, and thiols for complete enzymatic activity. Protein levels of Cathepsin C are detected in a variety of tissues, and it is most highly expressed in spleen, kidney, cytotoxic lymphocytes and myeloid cells, where it localizes to the secretory granule compartment. Cathepsin C is initially synthesized as a proenzyme that is rapidly processed to generate two distinct chains that function together as the mature form of the enzyme.Specifications
P53634 | |
2 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CTSC | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
2 to 500 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
1075 | |
<12% | |
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 50 μL|Plasma, 50 μL|Supernatant, 50 μL | |
Cathepsin C | |
4 hr 45 min |