missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin E Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1294
This item is not returnable.
View return policy
Description
Cathepsin E Monoclonal specifically detects Cathepsin E in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cathepsin E | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
NP_001901 | |
CTSE | |
Mouse myeloma cell line NS0-derived recombinant human Cathepsin E Gln18-Pro396 Accession # NP_001901 | |
500 μg | |
Primary | |
Detects human Cathepsin E in direct ELISAs and Western blots. It recognizes both the pro and mature forms of recombinant human (rh) Cathepsin E. In Western blots, 100% cross-reactivity with recombinant mouse (rm) Cathepsin E and recombinant human (rh) BACE is observed under reducing conditions. Under non-reducing conditions, less than 1% cross-reactivity with rmCathepsin E and rhBACE-1 is observed. Less than 1% cross-reactivity with rhCathepsin D is observed under both reducing and non-reducing conditions. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
212211 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CATE, cathepsin E, CTSE, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1510 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
For Research Use Only