missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL2 (MCP-1) Recombinant Protein, PeproTech®

Recombinant Protein

£67.75 - £2510.00

Specifications

Accession Number P13500
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6347
Molecular Weight (g/mol) 8.6 kDa
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17811753
View Documents
Gibco™
300-04-100UG
100 μg
£502.00
100µg
Please to purchase this item. Need a web account? Register with us today!
17891673
View Documents Promotion Details
Gibco™
300-04-1MG
1 mg
£2510.00
1mg
Please to purchase this item. Need a web account? Register with us today!
17871043
View Documents
Gibco™
300-04-20UG
20 μg
£164.00
20µg
Please to purchase this item. Need a web account? Register with us today!
17811073
View Documents Promotion Details
Gibco™
300-04-250UG
250 μg
£876.00
250µg
Please to purchase this item. Need a web account? Register with us today!
17870383
View Documents Promotion Details
Gibco™
300-04-500UG
500 μg
£1500.00
500µg
Please to purchase this item. Need a web account? Register with us today!
17830213
View Documents
Gibco™
300-04-50UG
50 μg
£305.00
50µg
Please to purchase this item. Need a web account? Register with us today!
17860563
View Documents
Gibco™
300-04-5UG
5 μg
£67.75
5µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Recombinant Human MCP-1 is an 8.6 kDa protein containing 76 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
Specifications

Specifications

P13500
protein with no preservative
8.6 kDa
E. coli
<1 EU/ μg
CCL2 (MCP-1)
Determined by its ability to chemoattract human monocytes using a concentration range of 10.0-100.0 ng/ml
Recombinant
-20°C
Lyophilized
ELISA, Functional Assay, Western Blot
6347
Human CCL2 (MCP-1)
RUO
Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1 A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1 A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF
Ccl2
Unconjugated
QPDAINAPVT CCYNFTNRKI SVQRLASYRR ITSSKCPKEA VIFKTIVAKE ICADPKQKWV QDSMDHLDKQ TQTPKT
E. coli
≥ 98% by SDS-PAGE gel and HPLC analyses.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.