missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CCL25 (TECK), Animal-Free Recombinant Protein, PeproTech®
Recombinant Protein
£100.00 - £1147.00
Specifications
Accession Number | O15444 |
---|---|
For Use With (Application) | Functional Assay |
Format | Lyophilized |
Formulation | protein with no preservative |
Gene ID (Entrez) | 6370 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17806538
|
Gibco™
AF-300-45-5UG |
5 μg |
£100.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17836828
|
Gibco™
AF-300-45-250UG |
250 μg |
£1147.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemokines are likely to play an important role in regulating the trafficking of developing T cells within the thymus. Chemokine C-C thymus expressed chemokine (TECK), also designated chemokine ligand 25 (CCL25), small induc-ible cytokine A25, chemokine b-15 or CK b-15, is expressed predominantly in thymic dendritic cells, thymic epithelial cells and in the small intestine. TECK, a CCR9 ligand, has suppressive activity against immature subsets of myeloid progenitors which have been stimulated to proliferate by multiple growth factors. TECK delivers signals through CCR9, which is important for the nav-igation of developing thymocytes. Bone marrow pre-pro-B cells and cells capable of generating pro-B colonies in the presence of interleukin-7 and Flt-3 ligand migrate to TECK, a response lost in later stages of B cell development.Specifications
O15444 | |
Lyophilized | |
6370 | |
CCL25 | |
-20°C | |
<0.1 EU/ μg | |
CCL25 | |
Human | |
Recombinant | |
Determined by its ability to bind CCR9 receptor expressed on the surface of MOLT4 cells. | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
protein with no preservative | |
14.2 kDa | |
HPLC, SDS-PAGE | |
RUO | |
A130072A22Rik; AI852536; C-C motif chemokine 25; C-C motif chemokine ligand 25; CCL25; chemokine (C-C motif) ligand 25; chemokine TECK; Ck beta-15; CKb15; Scya25; SCYA25TECKvar; small inducible chemokine 25; small inducible cytokine A25; small inducible cytokine subfamily A (Cys-Cys), member 25; small-inducible cytokine A25; Teck; TECKMGC150327; TECKvar; thymus expressed chemokine; thymus-expressed chemokine | |
Protein | |
Unconjugated | |
QGVFEDCCLA YHYPIGWAVL RRAWTYRIQE VSGSCNLPAA IFYLPKRHRK VCGNPKSREV QRAMKLLDAR NKVFAKLHHN MQTFQAGPHA VKKLSSGNSK LSSSKFSNPI SSSKRNVSLL ISANSGL | |
E. coli |