missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR2 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
£192.00 - £477.00
Specifications
Antigen | CCR2 |
---|---|
Clone | 48607 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18737889
|
R&D Systems
FAB151N-025 |
25 Tests |
£192.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755377
|
R&D Systems
FAB151N-100 |
100 Tests |
£477.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR2 Monoclonal specifically detects CCR2 in Human samples. It is validated for Flow Cytometry.Specifications
CCR2 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
CCR2 | |
IgG2b | |
Protein A or G purified from ascites | |
Detects human CCR2 transfectants but not the parental cell line or CCR5 transfectants. |
48607 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Mouse | |
NP_001116868 | |
729230 | |
NS0 mouse myeloma cell line transfected with human CCR2 Met1-Leu360 Accession # NP_001116868 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human CCR2 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™