Learn More
Novus Biologicals™ Human CCR8 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-75116
Additional Details : Weight : 0.00970kg
Description
The Human CCR8 ELISA Kit (Colorimetric) measures human CCR8 in serum, plasma and other biological fluids.Specifications
0.16 - 10 ng/mL | |
This kit recognizes Human CCR8 in samples. No significant cross-reactivity or interference between Human CCR8 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1237 | |
Human | |
GPCR | |
CCR8 |
0.10 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CCR8 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |