missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD 59 ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EH76RBX10
Additional Details : Weight : 2.50000kg
Description
CD59 (Protectin) is a small (18-20 kDa) GPI-anchored ubiquitously expressed inhibitor of the membrane attack complex (MAC). CD59 is the key regulator that preserves the autologous cells from terminal effector mechanism of the complement cascade. CD59 also associates with C5b-8 complex and counteracts appropriate formation of cytolytic pore within the plasma membrane. Further, CD59 is a low-affinity ligand of human CD2, causes T cell costimulation, and is involved in lymphocyte signal transduction. CD59 is a potent inhibitor of the complement membrane attack complex, whereby it binds complement C8 and/or C9 during the assembly of this complex, thereby inhibiting the incorporation of multiple copies of C9 into the complex, which is necessary for osmolytic pore formation. Mutations in this gene cause CD59 deficiency, a disease involving hemolytic anemia, thrombosis, and cerebral infarction. Multiple alternatively spliced transcript variants of CD59 have been identified.Specifications
P13987 | |
40 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CD59 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
40 to 10,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
966 | |
<12% | |
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 50 μL|Plasma, 50 μL|Supernatant, 50 μL | |
CD59 | |
4 hr 45 min |