missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CD155/PVR ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EH79RBX10

Product Code. 17727853

  • £3950.00 / Pack of 10
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P15151
0.8 ng/mL
ELISA
Colorimetric Microplate Reader
PVR
<10%
HRP
RUO
2°C to 8°C
1 hr 20 min
0.8 to 200 ng/mL
Biotin
Plasma, Serum, Supernatant
5817
<12%
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers
10 x 96 Tests
Serum, 50 μL|Plasma, 50 μL|Supernatant, 50 μL
CD155
4 hr 45 min
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers