Learn More
Invitrogen™ Human CD253 (TRAIL) (aa 207-220) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP046
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human TRAIL (residues 207-220) Synthetic Peptide, PEP-046, from Invitrogen.
PEP-046 is a 15 amino acid synthetic peptide whose sequence corresponds to residues 207-220 of human TRAIL. The sequence of this peptide is (amino to carboxy terminus): C-V207-Q-Y-I-Y-K-Y-T-S-Y-P-D-P-I220 (The N-terminal cysteine has been added to facilitate conjugation and is not a part of the native human TRAIL sequence)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and TRAIL, catalog # PA1-955. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
8743 | |
purified | |
Human CD253 (TRAIL) (aa 207-220) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P50591 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
TNFSF10 | |
No preservative | |
50 μg | |
C - V207 - Q - Y - I - Y - K - Y - T - S - Y - P - D - P - I220 |