missing translation for 'onlineSavingsMsg'
Learn More

Proteintech Human CD38 ELISA Kit

Solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay Kit

Brand:  Proteintech KE00253-96T


Product Code. 17650107

This item is not returnable. View return policy

Description

Description

KE00253 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The CD38 ELISA kit is to be used to detect and quantify protein levels of endogenous CD38. The assay recognizes human CD38. An antibody specific for CD38 has been pre-coated onto the microwells. The CD38 protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody specific for CD38 is added to detect the captured CD38 protein. For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
Specifications

Specifications

0.029 ng/mL
Cell Lysates, Plasma, Serum
Sandwich ELISA
Human
Sandwich
96 Tests
1 x 96 well plate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Human CD38 ELISA Kit, 96 RXN

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers