Learn More
ACROBiosystems Human CD40 / TNFRSF5 Protein, Fc Tag
MALS verified
£302.41 - £2310.44
Specifications
Accession Number | NP_001241.1 |
---|---|
Molecular Weight (g/mol) | 45.3 kDa |
Source | Human CD40 Protein, Fc Tag (MALS verified) (CD0-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1). |
Immunogen | Glu 21 - Arg 193 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15696614
|
ACROBiosystems
CD0-H5253-100UG |
100 μg |
£302.41
100µg |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15856127
|
ACROBiosystems
CD0-H5253-1MG |
1 mg |
£2310.44
1mg |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD40 is also known as TNFRSF5, Bp50, CDW40, MGC9013, TNFRSF5 and p50, is a member of the TNF receptor superfamily which are single transmembrane-spanning glycoproteins, and plays an essential role in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. CD40 is a costimulatory protein found on antigen presenting cells and is required for their activation. The binding of CD154 (CD40L) on TH cells to CD40 activates antigen presenting cells and induces a variety of downstream effects. CD40 contains 4 cysteine-rich repeats in the extracellular domain, and is expressed in B cells, dendritic cells, macrophages, endothelial cells, and several tumor cell lines. The extracellular domain has the cysteinerich repeat regions, which are characteristic for many of the receptors of the TNF superfamily. Interaction of CD40 with its ligand, CD40L, leads to aggregation of CD40 molecules,which in turn interact with cytoplasmic components to initiate signaling pathways. Early studies on the CD40-CD40L system revealed its role in humoral immunity. Defects in CD40 result in hyper-IgM immunodeficiency type 3 (HIGM3), an autosomal recessive disorder characterized by an inability of B cells to undergo isotype switching, as well as an inability to mount an antibody-specific immune response, and a lack of germinal center formation.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001241.1 | |
Human CD40 Protein, Fc Tag (MALS verified) (CD0-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Arg 193 (Accession # P25942-1). | |
-20°C | |
CD40 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
45.3 kDa | |
Glu 21 - Arg 193 | |
CD40,Bp50,CDW40,MGC9013,TNFRSF5,p50 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |