missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CDK6 Partial ORF (NP_001250, 3 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001021-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16073815

  • £280.00 / 10µg
Estimated Shipment: 29-08-2024
to see stock.

Description

Description

The protein encoded by this gene is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This kinase is a catalytic subunit of the protein kinase complex that is important for cell cycle G1 phase progression and G1/S transition. The activity of this kinase first appears in mid-G1 phase, which is controlled by the regulatory subunits including D-type cyclins and members of INK4 family of CDK inhibitors. This kinase, as well as CDK4, has been shown to phosphorylate, and thus regulate the activity of, tumor suppressor protein Rb. [provided by RefSeq]

Sequence: KDGLCRADQQYECVAEIGEGAYGKVFKARDLKNGGRFVALKRVRVQTGEEGMPLSTIREVAVLRHLETFEHPNVVRLFDVCTVSRTDRETKLTLVFE
Specifications

Specifications

NP_001250
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KDGLCRADQQYECVAEIGEGAYGKVFKARDLKNGGRFVALKRVRVQTGEEGMPLSTIREVAVLRHLETFEHPNVVRLFDVCTVSRTDRETKLTLVFE
RUO
CDK6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1021
CDK6 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC59692/PLSTIRE/STQTL11
CDK6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers