missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human COCO Antibody, R&D Systems™
Mouse Monoclonal Antibody
£146.86 - £448.15
Specifications
Antigen | COCO/DAND5 |
---|---|
Clone | 322209 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785809
|
R&D Systems
MAB30471-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18138790
|
R&D Systems
MAB30471 |
500 μg |
£448.15
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COCO/DAND5 Monoclonal specifically detects COCO/DAND5 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
COCO/DAND5 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1 | |
DAND5 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
322209 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q8N907 | |
199699 | |
E. coli-derived recombinant human COCO Arg23-Ala189 Accession # Q8N907 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human COCO in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Activin A, rhBMP-4, rhDAN, or recombinant mouse COCO is observed. |
For Research Use Only