missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CXCL10/IP-10 Quantikine ELISA Kit
Human CXCL10/IP-10 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ SIP100
This item is not returnable.
View return policy
Description
The Human CXCL10/IP-10 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CXCL10/IP-10/CRG-2 in cell culture supernates (10 ul), serum (75 ul), edta plasma (75 ul), heparin plasma (75 ul), saliva (10 ul), urine (100 ul).Specifications
7.81 - 500pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human IP-10 | |
Human | |
3627 | |
Cell Culture Supernates (10uL),Serum (75uL),EDTA Plasma (75uL),Heparin Plasma (75uL),Saliva (10uL),Urine (100uL) | |
C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 |
4.46pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum, Urine | |
Sufficient materials to run ELISAs on at least six 96-well plates | |
ELISA | |
6 Plates | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |