missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CXCL12 (SDF-1a), Animal-Free Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£177.00 - £2635.00
Specifications
Accession Number | P48061 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 6387 |
Molecular Weight (g/mol) | 8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17893273
|
Gibco™
AF-300-28A-10UG |
10 μg |
£177.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17843383
|
Gibco™
AF-300-28A-100UG |
100 μg |
£548.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17873063
|
Gibco™
AF-300-28A-1MG |
1 mg |
£2635.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17833383
|
Gibco™
AF-300-28A-500UG |
500 μg |
£1582.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
P48061 | |
protein with no preservative | |
8 | |
E. coli | |
RUO | |
12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; Cxcl12; H-SDF-1-alpha; Intercrine reduced in hepatomas; IRH; Pbsf; pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SCYB12; SDF 1 A; SDF 1 alpha; SDF 1 A; SDF1; SDF-1; SDF1 alpha; SDF-1 gamma; SDF1A; SDF-1-alpha(3-67); SDF1B; SDF-1-beta(3-72); SDF1a; Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tpar1 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
6387 | |
Human CXCL12 (SDF-1 A), Animal-Free | |
-20°C | |
<0.1 EU/μg | |
CXCL12 | |
Unconjugated | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |