missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CXCL16 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
£64.75 - £2400.00
Specifications
Accession Number | Q9H2A7 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 58191 |
Molecular Weight (g/mol) | 10.1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17851993
|
Gibco™
300-55-100UG |
100 μg |
£480.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17802453
|
Gibco™
300-55-1MG |
1 mg |
£2400.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17872703
|
Gibco™
300-55-250UG |
250 μg |
£838.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17872743
|
Gibco™
300-55-25UG |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17832173
|
Gibco™
300-55-500UG |
500 μg |
£1436.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17882253
|
Gibco™
300-55-5UG |
5 μg |
£64.75
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL16 is a member of the CXC chemokine family and signals through the CXCR6 receptor. CXCL16 may play a role in attracting lymphocyte subsets during inflammation and may facilitate certain immune responses. The chemokine domain of CXCL16 contains six cysteine residues, including the four highly conserved cysteine residues characteristic of CXC chemokines. The CXCL16 gene codes for a 273 amino acid polypeptide, which includes a 29 amino acid cytoplasmic domain and transmembrane sequence containing approximately 20 amino acids. The extracellular portion of CXCL16 contains a chemokines domain and an extended C-terminal mucin-like stalk sequence. The extracellular domain contains 89 amino acid residues (86 a.a. residues for the murine homolog).Specifications
Q9H2A7 | |
protein with no preservative | |
10.1 | |
E. coli | |
RUO | |
0910001K24Rik; AV290116; b2b498Clo; BB024863; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 16; CXC; Cxc chemokine ligand 16; CXC motif chemokine; C-X-C motif chemokine 16; C-X-C motif chemokine ligand 16; CXCL; Cxcl16; CXCL16v1; CXCL16v2; CXCLG16; Mutant line 498; scavenger receptor for phosphatidylserine and oxidized low density lipoprotein; SCYB16; Small-inducible cytokine B16; Srpsox; SR-PSOX; SR-PSOX/CXCL16; Transmembrane chemokine CXCL16; UNQ2759/PRO6714; zinc finger, MYND-type containing 15; Zmynd15 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
58191 | |
Human CXCL16 | |
-20°C | |
<1 EU/μg | |
Cxcl16 | |
Unconjugated | |
NEGSVTGSCY CGKRISSDSP PSVQFMNRLR KHLRAYHRCL YYTRFQLLSW SVCGGNKDPW VQELMSCLDL KECGHAYSGI VAHQKHLLP | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |