missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Cystatin B Quantikine ELISA Kit
a 4.5 hour solid phase ELISA designed to measure Cystatin B levels in cell culture supernates, serum, plasma, saliva, urine, and human milk.
Brand: R&D Systems™ DCYB00
Additional Details : Weight : 0.58200kg
Description
The Human Cystatin B Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Cystatin B/Stefin B in cell culture supernates (50 ul), serum (17 ul), edta plasma (17 ul), heparin plasma (17 ul), saliva (10 ul), urine (13 ul), human milk (10 ul).Specifications
0.16 - 10ng/mL | |
HRP | |
Natural and Recombinant Human Cystatin B | |
1476 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
0.034ng/mL | |
Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine | |
ELISA | |
Cell Culture Supernates (50uL),Serum (17uL),EDTA Plasma (17uL),Heparin Plasma (17uL),Saliva (10uL),Urine (13uL),Human Milk (10uL) | |
CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10 STFBcystatin-B, CSTB, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B |