Learn More
Gibco™ Human Dendritic Cell Cytokine Package, PeproTech®
Brand:  Gibco™ HDC
Product Code. 17813703
  • £696.00 / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Human Dendritic Cell Cytokine Package contains the key components required for the culturing of human dendritic cells.
IL-4 is a pleiotropic cytokine that regulates diverse T and B cell responses including cell proliferation, survival, and gene expression. Produced by mast cells, T cells, and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells.
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Functional Assay
Dendritic Cell Cytokine Package
Human
100 μg Human IL-4 (Cat. No. 200-04-100 UG), 100 μg Human GM-CSF (Cat. No. 300-03-100 UG)
1 Kit