missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Desmocollin-3 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4630-SP
This item is not returnable.
View return policy
Description
Desmocollin-3 Polyclonal specifically detects Desmocollin-3 in Human samples. It is validated for Western Blot.Specifications
Desmocollin-3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cadherin family member 3, CDHF3, CDHF3HT-CP, desmocollin 3, Desmocollin3, desmocollin-4, DSC, DSC1, DSC2, DSC3, DSC4, DSC4desmocollin-3, HT-CP | |
Sheep | |
Affinity Purified | |
RUO | |
1825 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q14574 | |
DSC3 | |
Mouse myeloma cell line NS0-derived recombinant human Desmocollin-3 Arg136-Lys690 Accession # Q14574 | |
25 μg | |
Primary | |
Detects human Desmocollin-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Desmocollin-2 is observed and less than 1% cross-reactivity with rhDesmocollin-1 is observed. | |
Human | |
IgG |
For Research Use Only