missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Desmocollin-3 Antibody, R&D Systems™
Sheep Polyclonal Antibody
£147.94 - £418.32
Specifications
Antigen | Desmocollin-3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18799077
|
R&D Systems
AF4630-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18028561
|
R&D Systems
AF4630 |
100 μg |
£418.32
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desmocollin-3 Polyclonal specifically detects Desmocollin-3 in Human samples. It is validated for Western Blot.Specifications
Desmocollin-3 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
Cadherin family member 3, CDHF3, CDHF3HT-CP, desmocollin 3, Desmocollin3, desmocollin-4, DSC, DSC1, DSC2, DSC3, DSC4, DSC4desmocollin-3, HT-CP | |
DSC3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q14574 | |
1825 | |
Mouse myeloma cell line NS0-derived recombinant human Desmocollin-3 Arg136-Lys690 Accession # Q14574 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Desmocollin-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Desmocollin-2 is observed and less than 1% cross-reactivity with rhDesmocollin-1 is observed. |
For Research Use Only