missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DRAM Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0456

Product Code. 15226669

  • £112.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

DRAM Synthetic Peptide, PEP-0456, from Invitrogen.

This peptide corresponds to 16 amino acid peptide from near the amino terminus of human DRAMPEP-0456 can be used as a blocking peptide with polyclonal antibody PA5-20336.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

55332
purified
Human DRAM
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q8N682
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
DRAM1
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.