missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human EGF, AOF Recombinant Protein
Human EGF, AOF Recombinant Protein
Brand: Gibco™ PHG6045
This item is not returnable.
View return policy
Description
Human EGF, AOF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
EGF has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse BALB/3T3 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
500 ug | |
Human | |
Carrier-Free | |
proliferation assay using balb/c 3T3 cells | |
500 μg | |
E. coli | |
Recombinant, Ligand | |
EGF (Epidermal Growth Factor) | |
Gibco™ |
6.2 kDa | |
EGF AOF | |
Store in freezer (-5 to -30°C). | |
0.1 ng/ml | |
< 1 EU/ml | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Stem Cell Research, Oncology | |
98 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.