Learn More
R&D Systems™ Human EMAP-II (NP_001135887.1) VersaClone cDNA
Brand: R&D Systems™ RDC3570
Additional Details : Weight : 0.00970kg
Specifications
EMAP-II/AIMP1 | |
9255 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_001135887.1 | |
AIMP1, aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPII, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43, SCYE1 | |
Human |