missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human EN-RAGE DuoSet ELISA
Human EN-RAGE DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DY1052-05
Includes: Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Description
The Human EN-RAGE DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human EN-RAGE/S100A12.Specifications
1 Kit (for 5 Plates) | |
Biotin | |
ELISA | |
CAAF1, CAAF1Neutrophil S100 protein, CAAFI, CAGC, CAGCS100, CAGCS100 calcium binding protein A12 (calgranulin C), Calcium-binding protein in amniotic fluid 1, Calgranulin C, Calgranulin-C, CGRPEN-RAGE, ENRAGE, Extracellular newly identified RAGE-binding protein, MRP6, p6calgranulin C, protein S100-A12, S100 calcium binding protein A12, S100 calcium-binding protein A12, S100 calcium-binding protein A12 (calgranulin C), S100A12 |
Human | |
EN-RAGE/S100A12 | |
6283 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |