missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human ENA-78 (CXCL5) (8-78 aa) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 300-22B-100UG
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation.Specifications
P42830 | |
protein with no preservative | |
7.8 | |
100 μg | |
-20°C | |
<1 EU/μg | |
CXCL5 | |
Unconjugated | |
LRELRCVCLQ TTQGVHPKMI SNLQVFAIGP QCSKVEVVAS LKNGKEICLD PEAPFLKKVI QKILDGGNKE N | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
6374 | |
Human ENA-78 (CXCL5) (8-78 aa) | |
E. coli | |
RUO | |
AMCF-II; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine ligand 5; CXCL; CXCL5; Cxcl6; Cytokine LIX; ENA78; ENA-78; ENA-78(1-78); ENA-78(8-78); ENA-78(9-78); Epithelial-derived neutrophil-activating protein 78; GCP-2; GCP-2(1-78); GCP-2(9-78); granulocyte chemotactic protein-2; LIX; Neutrophil-activating peptide ENA-78; neutrophil-activating protein 78; Scyb5; Scyb6; small inducible cytokine B subfamily, member 5; small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derived neutrophil-activating peptide 78); small inducible cytokine subfamily B, member 15; small-inducible cytokine B5 | |
Human | |
Recombinant | |
Lyophilized |