Learn More
Gibco™ Human EPO (Erythropoietin) Recombinant Protein
Description
Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in sterile, deionized water to a concentration of 12,000-120,000 IU/mL. Further dilutions can be made in aqueous buffers. For long term storage, EPO solutions should be supplemented with carrier protein, such as 0.1% HSA or BSA. Storage: This lyophilized preparation is stable at room temperature for three weeks, but should be stored desiccated below -18°C. Upon reconstitution, the preparation is stable for up to one week at 2°C to 8°C, but should be stored below -18°C for long term storage. Avoid repeated freeze/thaw cycles.
Specifications
Specifications
| Accession Number | P01588 |
| For Use With (Application) | Bioactivity |
| Formulation | 0.006mg citric acid, 0.59mg sodium citrate with 0.58mg NaCl and no preservative |
| Gene ID (Entrez) | 2056 |
| Molecular Weight (g/mol) | 30.4 kDa |
| Name | Human EPO (Erythropoietin) |
| Purification Method | Purified |
| Quantity | 500 IU |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.