missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human EPO (Erythropoietin) Recombinant Protein, PeproTech®

CHO cells Recombinant Protein

£66.00 - £1222.00

Specifications

Accession Number P01588
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 2056
Molecular Weight (g/mol) 18.4
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17830773
View Documents
Gibco™
100-64-100UG
100 μg
£245.00
100µg
Please to purchase this item. Need a web account? Register with us today!
17831643
View Documents
Gibco™
100-64-10UG
10 μg
£66.00
10µg
Please to purchase this item. Need a web account? Register with us today!
17810463
View Documents
Gibco™
100-64-1MG
1 mg
£1222.00
1mg
Please to purchase this item. Need a web account? Register with us today!
17871643
View Documents
Gibco™
100-64-250UG
250 μg
£429.00
250µg
Please to purchase this item. Need a web account? Register with us today!
17850553
View Documents
Gibco™
100-64-500UG
500 μg
£734.00
500µg
Please to purchase this item. Need a web account? Register with us today!
17801043
View Documents
Gibco™
100-64-50UG
50 μg
£160.00
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.
Specifications

Specifications

P01588
protein with no preservative
18.4
CHO cells
RUO
EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2
Human
Recombinant
Lyophilized
ELISA, Functional Assay, Western Blot
2056
Human EPO (Erythropoietin)
-20°C
<1 EU/μg
EPO
Unconjugated
APPRLICDSR VLERYLLEAK EAENITTGCA EHCSLNENIT VPDTKVNFYA WKRMEVGQQA VEVWQGLALL SEAVLRGQAL LVNSSQPWEP LQLHVDKAVS GLRSLTTLLR ALGAQKEAIS PPDAASAAPL RTITADTFRK LFRVYSNFLR GKLKLYTGEA CRTGDR
≥ 90% by SDS-PAGE gel and HPLC analyses.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.