missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Exostosin-like 3/EXTL3 Lumenal Domain Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
£147.94 - £418.32
Specifications
Antigen | Exostosin-like 3/EXTL3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18770808
|
R&D Systems
AF2635-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18020712
|
R&D Systems
AF2635 |
100 μg |
£418.32
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Exostosin-like 3/EXTL3 Polyclonal specifically detects Exostosin-like 3/EXTL3 in Human samples. It is validated for Western Blot.Specifications
Exostosin-like 3/EXTL3 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
botv, DKFZp686C2342, EC 2.4.1.223, exostoses (multiple)-like 3, Exostosin like 3, exostosin-like 3, EXTL1L, EXTL3, EXTR1BOTV, EXT-related protein 1, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, Hereditary multiple exostoses gene isolog, KIAA0519, Multiple exostosis-like protein 3, Putative tumor suppressor protein EXTL3, REG, reg receptor, REGR, RPR | |
EXTL3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O43909 | |
2137 | |
Mouse myeloma cell line NS0-derived recombinant human Exostosin-like 3/EXTL3 Lumenal Domain Thr52-Ile919 Accession # O43909 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Exostosin-like 3/EXTL3 Lumenal Domain in direct ELISAs and Western blots. |
For Research Use Only