missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EZH1 Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP1251

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15285249

  • £112.00 / 50µg
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

EZH1 Synthetic Peptide, PEP-1251, from Invitrogen.

This peptide corresponds to 18 amino acids near the amino terminus of human EZH1.PEP-1251 can be used as a blocking peptide with polyclonal antibody PA5-21137.
Specifications

Specifications

2145
purified
Human EZH1
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q92800
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
EZH1
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers